Learn More
Jackson Immuno Research Labs Peroxidase Streptavidin 1.0mg
SDP
Supplier:  Jackson Immuno Research Labs 016030084
Encompass_Preferred
Steptavidin, a bacterial protein isolated from Streptomyces avidinii, is similar to egg-white avidin in its ability to bind biotin, and has been used as a replacement for egg-white avidin because of its more favorable chemical properties.Conjugates of streptavidin are recommended for use with Biotin-SP-conjugated antibodies and Biotin-SP-conjugated ChromPure proteins. Buffer: 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 Stabilizer: 15 mg/ml Bovine Serum Albumin (IgG-Free, Protease-Free) Preservative: None (Warning: Use of sodium azide as a preservative will substantially inhibit the enzyme activity of horseradish peroxidase.)
Catalog No. NC9705430
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov