Learn More
Crescent Chemical Co Inc STREPTAVIDIN LYOPHIL SALT-FREE
SDP
Supplier:  Crescent Chemical Co Inc SE35490.02
Encompass_Preferred
Streptavidin lyophil. salt-free Avidin from Streptomyces avidinii, isolated from fermentation filtrates by ion exchange chromatography. Size - 5MG Storage Conditions - -15 °C TO -25 °C Catalog Number - 35490.02 CAS 9013-20-1
Catalog No. 50-248-160