Learn More
New England Biolabs, Inc. Streptavidin Magnetic Beads – 5 ml
SDP
Catalog No. 50812660
Encompass
Streptavidin Magnetic Beads are 1 micron superparamagnetic particles covalently coupled to a highly pure form of streptavidin. The beads can be used to capture biotin labeled substrates including antigens, antibodies and nucleic acids.
Catalog No. 50-812-660 Supplier New England Biolabs, Inc. Supplier No. S1420S
Streptavidin Magnetic Beads are 1 micron superparamagnetic particles covalently coupled to a highly pure form of streptavidin. The beads can be used to capture biotin labeled substrates including antigens, antibodies and nucleic acids.