Learn More
Sigma Aldrich Fine Chemicals Biosciences Streptavidin Mag Sepharose
Catalog No. 502904247
Encompass
Streptavidin Mag Sepharose(R) is a magnetic bead for simple and efficient enrichment of target proteins by immunoprecipitation and purification of biotinylated biomolecules. Streptavidin Mag Sepharose(R) utilizes the strong interaction between biotin and streptavidin ligand which is immobilized on magnetic beads. Magnetic beads simplify sample handling in small-scale purifications.
Catalog No. 50-290-4247 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28985738
Streptavidin Mag Sepharose(R) is a magnetic bead for simple and efficient enrichment of target proteins by immunoprecipitation and purification of biotinylated biomolecules. Streptavidin Mag Sepharose(R) utilizes the strong interaction between biotin and streptavidin ligand which is immobilized on magnetic beads. Magnetic beads simplify sample handling in small-scale purifications.