Learn More
Vector Laboratories MagnaLINK Streptavidin Magnetic Beads 1ml
SDP
Catalog No. 502045731
Encompass
MagnaLINK 2.8 µm Streptavidin Magnetic Beads offer the highest biotin binding capacity on the market, combined with exceptional size uniformity. Higher binding translates to reduced bead mass required to immobilize a biotinylated sample and lower background from nonspecific binding, resulting in better signals and lower costs.
Catalog No. 50-204-5731 Supplier Vector Laboratories Supplier No. M1003010
MagnaLINK 2.8 µm Streptavidin Magnetic Beads offer the highest biotin binding capacity on the market, combined with exceptional size uniformity. Higher binding translates to reduced bead mass required to immobilize a biotinylated sample and lower background from nonspecific binding, resulting in better signals and lower costs.