Learn More
Genscript Corporation Streptavidin MagBeads
Supplier:  Genscript Corporation L00936
Encompass
GenScript Streptavidin MagBeads are ideal for fast and convenient purification of biotinylated proteins peptides antibodies and oligonucleotides and protein interaction studies
Catalog No. 50-000-57395