Learn More
LUNA NANOTECH INC Streptavidin Magnetic Beads, 2 ML
Catalog No. 502566972
Encompass_Preferred
Streptavidin Coated Magnetic Beads, 500 nm Diameter. 2 mL @ 10 mg/mL
500 nm diameter magnetic beads with Streptavdin protein functionalized surface.
Features:
- High binding capacity of Biotin-conjugated molecules:
- 2,000 to 4,000 pmole of biotin / mg of magnetic beads
- 15-20 µg botinylated antibody / mg of magnetic beads
- Rapid and efficient biomolecule purification from complex samples.
- Fast response to a magnet.
- High monodispersity.
Applications:
Magnetic separation of Biotinylated cells, DNA, proteins, antibodies, and small ligands from complex samples. The streptavidin-coated magnetic beads are simply added to Biotinylated molecules for binding. The samples are transferred into a magnetic rack for easy removal of unbound residue.
Citations:
J. H. Molinski, S. Parwal and J. X. J. Zhang, "Laser-Manufactured
Catalog No. 50-256-6972 Supplier LUNA NANOTECH INC Supplier No. MGBSTRP102
Streptavidin Coated Magnetic Beads, 500 nm Diameter. 2 mL @ 10 mg/mL
500 nm diameter magnetic beads with Streptavdin protein functionalized surface.
Features:
- High binding capacity of Biotin-conjugated molecules:
- 2,000 to 4,000 pmole of biotin / mg of magnetic beads
- 15-20 µg botinylated antibody / mg of magnetic beads
- Rapid and efficient biomolecule purification from complex samples.
- Fast response to a magnet.
- High monodispersity.
Applications:
Magnetic separation of Biotinylated cells, DNA, proteins, antibodies, and small ligands from complex samples. The streptavidin-coated magnetic beads are simply added to Biotinylated molecules for binding. The samples are transferred into a magnetic rack for easy removal of unbound residue.
Citations:
J. H. Molinski, S. Parwal and J. X. J. Zhang, "Laser-Manufactured