Learn More
G-Biosciences Streptavidin Magnetic Beads

Description
Biotinylated molecules, such as nucleic acids, peptides and proteins can bind to Streptavidin Magnetic Beads easily. By applying magnetic attraction, Beads-molecules complex will be temporarily immobilized at tube wall, so the other parts in supernatant can be removed easily and efficiently.
Specification:
- Core: Iron Oxide (Fe3O4)
- Layer: Dextran
- Surface protein: Streptavidin
- Mean Diameter of Particles: ∼1 μm
- Storage Buffer: PBS pH 7.4, 0.02% Tween 20, 0.09% sodium azide and 10% glycerol
- Storage temperature: 2 to 8°C
- Capacity: 12 nmole biotin/ mL
Specifications
Specifications
| Particle Size | 1 to 3 μm |
| Form | Resin Slurry |
| Product Line | DynaMag |
| Quantity | 5 mL |
| For Use With (Application) | Immunoprecipitation, purification and other applications. |
| Type | Magnetic Bead Protein Purification System |
| Capacity (Metric) | 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.