Learn More
Bioworld STREPTAVIDIN- NANO-ACTIVEGOLD
Supplier:  Bioworld 201000112
Encompass_Preferred
Nano-activeGold is a newly developed gold immunoprobe. Nano-ActiveGold reagent consist ofStreptavidin covalently conjugated to the 1.5 nm Nano-activeGold cluster label. Nano-activeGold can be used in immunoblotting, light microscopy, an...
Catalog No. 50-197-486