Learn More
Spherotech STREPTAVIDIN PARTICLES
Catalog No. NC0831215
Encompass_Preferred
Streptavidin Polystyrene Particles, 0.5% w/v, 3.0-3.4 um, 5 mL
Catalog No. NC0831215 Supplier Spherotech Supplier No. SVP305
Streptavidin Polystyrene Particles, 0.5% w/v, 3.0-3.4 um, 5 mL