Learn More
MOSS INC Streptavidin-PE conjugate, 1mL
Catalog No. NC0609463
Encompass_Preferred
MOSS Specialized Conjugates are produced in Homogeneous, Liquid Stable form in batches ranging from 10mgs to >1000mgs. They are suitable for use on various Immunoassay, Flow Cytometry and Multiplexing platforms such as Luminex*.  MOSS conjugates produce exceptional signal-to-noise ratios and can be customized for maximum performance.
Catalog No. NC0609463 Supplier MOSS INC Supplier No. SAPE001
MOSS Specialized Conjugates are produced in Homogeneous, Liquid Stable form in batches ranging from 10mgs to >1000mgs. They are suitable for use on various Immunoassay, Flow Cytometry and Multiplexing platforms such as Luminex*.  MOSS conjugates produce exceptional signal-to-noise ratios and can be customized for maximum performance.