Learn More
Rockland Immunochemicals STREPTAVIDIN PEROXIDASE Conjugated - S000-03
SDP
Supplier:  Rockland Immunochemicals S00003
Encompass
Streptavidin Peroxidase is a useful detection reagent for primary antibodies conjugated to biotin. Streptavidin Peroxidase can be utilized in Immunohistochemistry, Western Blotting, and ELISA experiment formats in combination with the proper substrate (TMB-1000 or FEMTOMAX-110).
Catalog No. 50-105-9144