Learn More
Biogenex Laboratories Streptavidin Peroxidase Diluent
SDP
Catalog No. NC9718958
Encompass_Preferred
The Common Antibody Diluent and Enhanced Antibody Diluents have been developed for use with all antibodies in immunohistochemistry and have been specifically optimized for use with BioGenex antibodies and reagent products.
Catalog No. NC9718958 Supplier Biogenex Laboratories Supplier No. HK1575K
The Common Antibody Diluent and Enhanced Antibody Diluents have been developed for use with all antibodies in immunohistochemistry and have been specifically optimized for use with BioGenex antibodies and reagent products.