Learn More
Sigma Aldrich Fine Chemicals Biosciences Streptavidin Peroxidase fr2MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S55122MG
Encompass
Streptavidin is considered as a high-affinity biotin-binding agent which shows resistance to extreme pH detergents temperature denaturants and enzymes hence it is used in molecular biology and bionanotechnology.
Catalog No. 50-290-4903