Learn More
Description
Streptavidin for Western Blot, ICC/IF, IHC, IP, ELISA
Specifications
Specifications
| Concentration | 1mg/mL |
| Conjugate | DyLight 488 |
| Cross Reactivity | Tag |
| Formulation | PBS with 1% BSA and 0.02% sodium azide; pH 7.2 |
| pH | 7.2 |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Content And Storage | Stable for 1 year at 4°C |
| Form | Liquid |
| For Use With (Application) | ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.