Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Streptavidin Protein, DyLight™ 650

Catalog No. 84547 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-547 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 84-547 Supplier Invitrogen™ Supplier No. 84547
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Protein

Streptavidin for ICC/IF

Streptavidin is a tetrameric protein purified from Streptomyces avidinii that binds very tightly to the vitamin biotin with a Kd of ~ 10-14 mol/l. The high affinity recognition of biotin and biotinylated molecules has made streptavidin one of the most important components in diagnostics and laboratory kits.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL, 1mg/mL
For Use With (Application) Immunocytochemistry, Immunofluorescence
Formulation Buffered saline solution with 1% BSA and no preservative
Name Streptavidin
Conjugate DyLight 650
Cross Reactivity Tag
Purification Method Purified
Quantity 1 mL
Storage Requirements 4°C
Regulatory Status RUO
Common Name Streptavidin
Product Type Protein, Streptavidin
Content And Storage 4°C, Stable for 1 year at 4°C
Form Liquid
Purity or Quality Grade purified
Host Species N/A
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.