Learn More
ACROBiosystems Streptavidin Protein, Tb³± Chelate, TR-FRET, 20000 Tests
Catalog No. 5000807410
Encompass_Preferred
This streptavidin protein is a universal tool for TR-FRET assays designed to bind biotinylated molecules. It consists of high-purity streptavidin covalently conjugated with Tb3+ chelate. When used in combination with acceptor-labeled molecules, it facilitates a FRET reaction upon excitation by a specific light source, resulting in a 665 nm emission signal when molecules are within 10 nm proximity.
  • Expressed from E. coli cells
  • Supplied as lyophilized powder from 0.22 μm filtered solution in PBS, pH 7.4 with trehalose as protectant
  • Validated for binding biotinylated molecules as an acceptor in TR-FRET assays
  • Requires reconstitution according to the specific certificate of analysis protocol
Catalog No. 50-008-07410 Supplier ACROBiosystems Supplier No. FRTTA0120000TESTS
This streptavidin protein is a universal tool for TR-FRET assays designed to bind biotinylated molecules. It consists of high-purity streptavidin covalently conjugated with Tb3+ chelate. When used in combination with acceptor-labeled molecules, it facilitates a FRET reaction upon excitation by a specific light source, resulting in a 665 nm emission signal when molecules are within 10 nm proximity.
  • Expressed from E. coli cells
  • Supplied as lyophilized powder from 0.22 μm filtered solution in PBS, pH 7.4 with trehalose as protectant
  • Validated for binding biotinylated molecules as an acceptor in TR-FRET assays
  • Requires reconstitution according to the specific certificate of analysis protocol