Learn More
Description
Thermo Scientific™ Pierce™ Streptavidin, Horseradish Peroxidase Conjugate includes streptavidin in a purified form conjugated to peroxidase for substrate-based detection.
Thermo Scientific™ Pierce™ Streptavidin, Horseradish Peroxidase Conjugate includes streptavidin in a purified form conjugated to peroxidase for substrate-based detection.
Streptavidin, Horseradish Peroxidase Conjugate enables detection of biotinylated antibodies and other probes in a variety of standard assay methods, including western blotting, ELISA, immunohistochemistry, and fluorescence imaging. Streptavidin, Horseradish Peroxidase conjugate is supplied as lyophilized powder in phosphate-based buffer for immediate reconstitution with water.
Streptavidin is biotin-binding protein that was originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Invitrogen Streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation. This product has been successfully used in ELISA, Immunohistochemistry and Western Blot procedures.
Specifications
Specifications
| Conjugate | HRP |
| Quantity | 2 mg |
| Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C. |
| Shipping Condition | Ambient Temperature |
| Form | Solid |
| Product Line | Pierce |
| Product Type | Streptavidin Conjugate |
| Target Molecule | Biotin |
| Label or Dye | HRP (Horseradish Peroxidase) |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.