Learn More
Leinco Technologies Inc Streptavidin Purified | 5mg
SDP
Catalog No. NC1028410
Encompass_Preferred
Streptavidin is a biotin binding protein present in the fermentation broth of the bacterium Streptomyces avidinii. Each molecule of streptavidin can bind four molecules of biotin with a high affinity constant (Kd~10 -15). Unlike native avidin, streptavidin is not glycosylated and has a near neutral isoelectric point (pI ~ 5-6) vs a pI of 10 for native avidin.
Catalog No. NC1028410 Supplier Leinco Technologies Inc Supplier No. S2035MG
Streptavidin is a biotin binding protein present in the fermentation broth of the bacterium Streptomyces avidinii. Each molecule of streptavidin can bind four molecules of biotin with a high affinity constant (Kd~10 -15). Unlike native avidin, streptavidin is not glycosylated and has a near neutral isoelectric point (pI ~ 5-6) vs a pI of 10 for native avidin.