Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Streptavidin, (R-PE)

Catalog No. SA10041
Encompass_Preferred
Change view
Click to view available options
Quantity:
0.5 mL
2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SA10041 0.5 mL
50-112-1548 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SA10041 Supplier Invitrogen™ Supplier No. SA10041
Add to Cart
Add to Cart

Streptavidin, (R-PE)

  • Class: RUO
  • Statement: For research use only. Not for use in diagnostic procedures.
  • Buffer: PBS
  • Preservative: 0.1% Sodium Azide
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
Conjugate R-phycoerythrin
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis
Quantity 0.5 mL
Content And Storage Contains 1 vial of streptavidin,R-PE conjugate.

Store in refrigerator (2–8°C).
Excitation/Emission 488/575
Flow Cytometer Laser Lines 488, 488 or 561, 561
Shipping Condition Wet Ice
Form Solution
Buffer PBS Buffer
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye R-PE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.