Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Streptavidin, R-Phycoerythrin Conjugate (SAPE) - premium grade - 1 mg/mL

Catalog No. S21388
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S21388 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S21388 Supplier Invitrogen™ Supplier No. S21388
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Streptavidin, R-Phycoerythrin Conjugate (SAPE) - premium grade - 1 mg/mL

The R-phycoerythrin (R-PE) conjugate of streptavidin comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (R-phycoerythrin). Streptavidin has a very high binding affinity for biotin, and a conjugate of streptavidin is commonly used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids, and other molecules (for example, a biotinylated primary antibody bound to a protein target can be detected with a fluorescently labeled streptavidin). Streptavidin R-PE is very valuable in biotin-streptavidin detection protocols developed for flow cytometry, microplate assays, and microarrays. This R-PE streptavidin conjugate is a high-purity product (purity confirmed by HPLC) and is supplied as 1 mL of a 1 mg/mL solution.

Important Features of Streptavidin RPE:
  • Ex/Em maxima of ∼ (496, 546, 565/578) nm
  • 5 to 10 times brighter than organic dyes
  • Very high solubility in aqueous solutions
  • Free of all unconjugated streptavidin; and mostly free of unconjugated R-PE
  • Ideal for flow cytometry, microplate assays, and microarrays

    Properties of R-phycoerythrin
    R-PE has a quantum efficiency of 78%, making it an incredibly bright reagent, useful where high sensitivity is needed. Because of poor photostability, however, it is not recommended for fluorescence microscopy.

    Find Biotinylated Conjugates
    We offer an extensive line of biotinylated conjugates for use in biotin-streptavidin detection strategies.
  • Use the primary antibody search tool to find biotinylated primary antibodies
  • Use the secondary antibody selector tool to find biotinylated secondary antibodies and biotinylated anti-dye and anti-hapten antibodies

    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

    Related Links:

    Learn more about Avidin-Biotin Detection

    Learn more about the practical applications for Streptavidin R-PE
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 1 mg/mL
    Conjugate R-phycoerythrin
    Quantity 1 mL
    Content And Storage Store in refrigerator (2–8°C) and protect from light.
    Shipping Condition Wet Ice
    Form Liquid
    Product Type Streptavidin Conjugate (fluorescent)
    WARNING: Cancer - www.P65Warnings.ca.gov

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.