Learn More
Bioworld Streptavidin (Recombinant), 5 mg
Supplier:  Bioworld 220607001
Encompass_Preferred
Streptavidin Specifications:From Streptomyces avidini Biotechnology GradeActivity: >17U/mg proteinElectrophoresis: Pass DNase and RNase Activity: None detectedProtease: None detectedProduct for in vitro and lab research use only, not for human use or consumption.
Catalog No. 50-253-800