Learn More
Sartorius Biosensor / Streptavidin (SA) Pack
Supplier:  Sartorius 185020
Encompass
Streptavidin (SA) Biosensors - Five trays of 96 biosensors coated with streptavidin for quantitation, screening, or kinetic applications
Catalog No. NC9825342
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov