Learn More
Bioworld STREPTAVIDIN SEPAROPOREAG
Supplier:  Bioworld 201810372
Encompass
STREPTAVIDIN SEPAROPORE (AGAROSE): Streptavidin Separopore Affinity matrix with immobilized Streptavidin by CNBr activation coupling method.; For purification of biotinylated compounds.; Supplied as buffered saline solution. Technical sp...
Catalog No. 50-197-758