Learn More
Bioworld STREPTAVIDIN SEPAROPOREAG
Catalog No. 50197757
Encompass
STREPTAVIDIN SEPAROPORE (AGAROSE): Streptavidin Separopore Affinity matrix with immobilized Streptavidin by CNBr activation coupling method.; For purification of biotinylated compounds.; Supplied as buffered saline solution. Technical sp...
Catalog No. 50-197-757 Supplier Bioworld Supplier No. 201810371
STREPTAVIDIN SEPAROPORE (AGAROSE): Streptavidin Separopore Affinity matrix with immobilized Streptavidin by CNBr activation coupling method.; For purification of biotinylated compounds.; Supplied as buffered saline solution. Technical sp...