Learn More
Apexbio Technology LLC STREPTAVIDIN-TEXAS-RED-X-1ML
SDP
Catalog No. 5000567451
Encompass_Preferred
Streptavidin-Texas red X is a fluorescent probe widely used in biomedical research particularly in the fields of cell biology molecular biology and immunology Originating as a conjugate of streptavidin and Texas red dye it operates by binding with high affinity to biotin This interaction facilitates the detection and quantification of biotinylated molecules within various assays In vitro this conjugate demonstrates high specificity making it ideal for use in flow cytometry fluorescence microscopy and ELISA Its fluorescence properties allow for sensitive detection in the nanomolar to low micromolar range Typical applications include staining of cells in culture imaging of animal tissues and use in drug screening protocols Experimental concentrations typically depend on the specific research design This versatile tool is instrumental for scientists aiming to visualize and analyze biological processes with precision and accuracy in diverse experimental setups
Catalog No. 50-005-67451 Supplier Apexbio Technology LLC Supplier No. K44071
Streptavidin-Texas red X is a fluorescent probe widely used in biomedical research particularly in the fields of cell biology molecular biology and immunology Originating as a conjugate of streptavidin and Texas red dye it operates by binding with high affinity to biotin This interaction facilitates the detection and quantification of biotinylated molecules within various assays In vitro this conjugate demonstrates high specificity making it ideal for use in flow cytometry fluorescence microscopy and ELISA Its fluorescence properties allow for sensitive detection in the nanomolar to low micromolar range Typical applications include staining of cells in culture imaging of animal tissues and use in drug screening protocols Experimental concentrations typically depend on the specific research design This versatile tool is instrumental for scientists aiming to visualize and analyze biological processes with precision and accuracy in diverse experimental setups