Learn More
Cayman Chemical StreptavIdInSureLIght RPE 1ea
SDP
Catalog No. 502955572
Encompass_Preferred
Streptavidin conjugated to SureLight R-PE Binding Moiety: Streptavidin Dye: SureLight R-PE Excitation max. λ: 565 > 540 > 498 nm Emission max. λ: 578 nm Uses: Flow cytometry, fluorescence-activated cell sorting, and luminex Important Features of SureLight™ Polymeric SAPE (high-intensity): • 8-10x brighter than most SAPE conjugates • 10-15% brighter and interchangeable with Moss’s SAPE 001,003 • 15-20nm hydrodynamic radius • Two year shelf-life • BSA-free • Competitive bulk pricing - call for details
Catalog No. 50-295-5572 Supplier Cayman Chemical Supplier No. 134341 EA
Streptavidin conjugated to SureLight R-PE Binding Moiety: Streptavidin Dye: SureLight R-PE Excitation max. λ: 565 > 540 > 498 nm Emission max. λ: 578 nm Uses: Flow cytometry, fluorescence-activated cell sorting, and luminex Important Features of SureLight™ Polymeric SAPE (high-intensity): • 8-10x brighter than most SAPE conjugates • 10-15% brighter and interchangeable with Moss’s SAPE 001,003 • 15-20nm hydrodynamic radius • Two year shelf-life • BSA-free • Competitive bulk pricing - call for details