Learn More
Sigma Aldrich Fine Chemicals Biosciences StreptaWellTM transparent
Catalog No. 502906054
Encompass
Transparent streptavidin-coated microplatesBlocked volume 300muLSA-coated area 300muL coating variance guaranteedWell geometry C
Catalog No. 50-290-6054 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11664778001
Transparent streptavidin-coated microplatesBlocked volume 300muLSA-coated area 300muL coating variance guaranteedWell geometry C