Learn More
Cayman Chemical StreptImIdon 2.5mg
SDP
Catalog No. 502989313
Encompass_Preferred
A bacterial metabolite with diverse biological activities; inhibits protein synthesis in a cell-free assay at 50 μg/ml; reduces tumor growth in H.S. No. 1 human sarcoma and H.Ep. No. 3 human epidermoid carcinoma rat xenograft models at 12.5 and 25 mg/kg per day
Catalog No. 50-298-9313 Supplier Cayman Chemical Supplier No. 299842.5 MG
Add to Cart
Add to Cart
A bacterial metabolite with diverse biological activities; inhibits protein synthesis in a cell-free assay at 50 μg/ml; reduces tumor growth in H.S. No. 1 human sarcoma and H.Ep. No. 3 human epidermoid carcinoma rat xenograft models at 12.5 and 25 mg/kg per day