Learn More
AdipoGen Streptochlorin (1 mg)
SDP
Supplier:  AdipoGen AGCN20141M001
Encompass_Preferred
Chemical. CAS: 120191-51-7. Formula: C11H7ClN2O. MW: 218.6. Isolated from Streptomyces roseolilacinus. Antibiotic. TNF-alpha-induced NF-kappaB activation inhibitor. Antiangiogenic. Apoptosis inducer. Anticancer compound. Selective cytotoxic against several cancer cell lines. Tyrosinase inhibitor.
Catalog No. 50-205-3519