Learn More
ATCC Streptococcus agalactiae
Supplier:  ATCC 13813
Encompass
Streptococcus agalactiae strain NCTC 8181 [G19] is a whole genome-sequenced strain that has applications in quality control and bioinformatics. This strain is non-hemolytic and is typed as Lancefield’s group B.
Catalog No. 50-238-2565