Learn More
ATCC Streptococcus agalactiae
Supplier:  ATCC 49446
Encompass
Streptococcus agalactiae strain 3139 is a whole-genome sequenced bacterium that was isolated in Jerusalem from a newborn with sepsis. This strain is typed as serotype IV.
Catalog No. 50-238-0849