Learn More
ATCC Streptococcus agalactiae; group B
Supplier:  ATCC 12401
Encompass
Streptococcus agalactiae typing strain H36B [ATCC 8057, J. Brown strain Baby B] is in Lancefield's group B, serotype Ib, and can be used as a quality control strain.
Catalog No. 50-238-2309