Learn More
ATCC Streptococcus agalactiae; O90R
Catalog No. 502381797
Encompass
Streptococcus agalactiae grouping strain O90R [E. Todd strain Aronson Wamoscher] is a whole-genome sequenced bacterial strain. The product is in Lancefield's group B and can be used as a control strain for Group B Streptococcus (GBS). It also has applications in media testing.
Catalog No. 50-238-1797 Supplier ATCC Supplier No. 12386
Streptococcus agalactiae grouping strain O90R [E. Todd strain Aronson Wamoscher] is a whole-genome sequenced bacterial strain. The product is in Lancefield's group B and can be used as a control strain for Group B Streptococcus (GBS). It also has applications in media testing.