Learn More
ATCC Streptococcus agalactiae; Strain: 5030-08
Catalog No. 502380977
Encompass
Streptococcus agalactiae strain 5030-08 is a Group B Streptococcus (GBS) that is sensitive to Ampicillin, Levofloxacin, Linezolid, Vancomycin, Tetracycline, and Tigecycline.
Catalog No. 50-238-0977 Supplier ATCC Supplier No. BAA2669
Streptococcus agalactiae strain 5030-08 is a Group B Streptococcus (GBS) that is sensitive to Ampicillin, Levofloxacin, Linezolid, Vancomycin, Tetracycline, and Tigecycline.