Learn More
ATCC Streptococcus agalactiae; Strain: 2010228816
Catalog No. 502381056
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Streptococcus agalactiae strain 2010228816 is a Group B Streptococcus (GBS) typed as serotype VI. This strain is sensitive to Ampicillin, Levofloxacin, Linezolid, Vancomycin, Tetracycline, and Tigecycline.
Catalog No. 50-238-1056 Supplier ATCC Supplier No. BAA2671
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Streptococcus agalactiae strain 2010228816 is a Group B Streptococcus (GBS) typed as serotype VI. This strain is sensitive to Ampicillin, Levofloxacin, Linezolid, Vancomycin, Tetracycline, and Tigecycline.