Learn More
ATCC Pseudomonas aeruginosa
Supplier:  ATCC 35032
Encompass
Strain of Pseudomonas aeruginosa with applications in opportunistic pathogen research and quality control testing. This strain is fluorescent and produces pyocyanin.
Catalog No. 50-238-0053