Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Streptococcus bovis Z167, Titered
SDP

Catalog No. 22158036 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-036 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-036 Supplier ZeptoMetrix Supplier No. 0804015
Add to Cart
Add to Cart

Used to determine limit of detection (LOD), in diagnostic assay development or crossreactivity studies

  • Each frozen aliquot contains 1mL of pure, titered culture of  Streptococcus bovis
  • Identification of this organism was confirmed by 16S sequencing
  • Purity of culture was monitored by Gram staining and by additional culturing
  • Titer was performed on one aliquot after freezing
  • Freezing medium contains 15% glycerol as cryoprotectant
  • Contain intact organisms and should be run in a manner identical to that used for clinical specimen

Specifications

Content And Storage 2°C to 8°C
Format Titered
Product Type Titered Live Bacteria Control
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.