Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 34 (d10A) 2mg
Catalog No. 502381456
Encompass
Purified pneumococcal polysaccharide type 10A is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 10A can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-1456 Supplier ATCC Supplier No. 509X
Purified pneumococcal polysaccharide type 10A is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 10A can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.