Learn More
ATCC Streptococcus equi; subsp. zooepidemicus
Catalog No. 502383522
Encompass
Streptococcus equi subsp. zooepidemicus strain NCDO 1358 [NCTC 4676, S34] was isolated in England from a case of bovine mastitis. This whole-genome sequenced bacterial type strain can be used in agricultural research and quality control.
Catalog No. 50-238-3522 Supplier ATCC Supplier No. 43079
Streptococcus equi subsp. zooepidemicus strain NCDO 1358 [NCTC 4676, S34] was isolated in England from a case of bovine mastitis. This whole-genome sequenced bacterial type strain can be used in agricultural research and quality control.