Learn More
ATCC Pasteurella multocida; subsp. multocida
Supplier:  ATCC 43137
Encompass
Pasteurella multocida subsp. multocida strain NCTC 10322 [W-9217] was isolated from a pig. This bacterial type strain is propagated aerobically in brain heart infusion medium.
Catalog No. 50-238-0491