Learn More
ATCC Streptococcus mutans
Catalog No. 502381768
Encompass
Streptococcus mutans strain NCTC 10449 [IFO 13955] is a whole-genome sequenced bacterial type strain isolated from carious dentine. This product has applications in media testing.
Catalog No. 50-238-1768 Supplier ATCC Supplier No. 25175
Streptococcus mutans strain NCTC 10449 [IFO 13955] is a whole-genome sequenced bacterial type strain isolated from carious dentine. This product has applications in media testing.