Learn More
ATCC Streptococcus mutans
Supplier:  ATCC 700610
Encompass
Streptococcus mutans strain UA159 [UAB577] is a bacterium that was isolated from a child with active caries. This culture is whole-genome sequenced and is a microbiome standard component.
Catalog No. 50-238-4899