Learn More
ATCC Klebsiella quasipneumoniae; 6 x 0.3mL
Catalog No. 502379573
Encompass
ATCC 700603-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 700603™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.
Catalog No. 50-237-9573 Supplier ATCC Supplier No. 700603MINIPACK
ATCC 700603-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 700603™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.