Learn More
ATCC Streptococcus pneumoniae
Supplier:  ATCC 6319
Encompass
Streptococcus pneumoniae is a bacterium with applications in respiratory disease research. This strain is typed as serotype 19 and exhibits optochin sensitivity.
Catalog No. 50-238-0030
 Restricted Item