Learn More
ATCC Mycobacterium avium
Supplier:  ATCC 700897
Encompass
Mycobacterium avium strain MAC 511 is a drug-resistant bacterium that was isolated in 1988 from human blood. This culture is proposed as a standard strain for MAC susceptibility testing.
Catalog No. 50-238-1117