Learn More
ATCC Streptococcus pneumoniae
Supplier:  ATCC BAA340
Encompass
MIC (ug/mL): penicillin 8; cefotaxime 8; erythromycin 8; clindamycin 4; chloramphenicol 2; tetracycline 1. Representative strain of the Tennessee (H1)14-18 pneumococcal clone as confirmed by the Pneumococcal Molecular Epidemiology Network (PMEN).
Catalog No. 50-238-1182
 Restricted Item