Learn More
ATCC Streptococcus pneumoniae; 262
Catalog No. 502384622
Encompass
Streptococcus pneumoniae strain 262 is a whole-genome sequenced bacterium that was isolated in Arizona from the sputum of a 75-year-old human male. This bacterial culture has applications in susceptibility disc testing and respiratory disease research. This strain is serotype 19 (Danish designation 19F).
Catalog No. 50-238-4622 Supplier ATCC Supplier No. 49619
Streptococcus pneumoniae strain 262 is a whole-genome sequenced bacterium that was isolated in Arizona from the sputum of a 75-year-old human male. This bacterial culture has applications in susceptibility disc testing and respiratory disease research. This strain is serotype 19 (Danish designation 19F).