Learn More
ATCC Streptococcus pneumoniae
Supplier:  ATCC BAA255
Encompass
Streptococcus pneumoniae strain R6 is a whole-genome sequenced bacterium that has applications in respiratory research. This strain is non-virulent.
Catalog No. 50-238-2560