Learn More
ATCC Streptococcus pneumoniae
Supplier:  ATCC 10366
Encompass
Streptococcus pneumoniae has applications in respiratory disease research. This bacterium is incubated in a 5% CO2 atmosphere to enhance growth.
Catalog No. 50-238-4814